Learn More
1-Methylpyrene, TRC
Specifications
Specifications
| Chemical Name or Material | 1-Methylpyrene |
| CAS | 2381-21-7 |
| Molecular Formula | C17 H12 |
| Quantity | 100 mg |
| Synonym | Pyrene, 1-methyl-, 1-Methylpyrene, 3-Methylpyrene, NSC 90776 |
| InChI Formula | InChI=1S/C17H12/c1-11-5-6-14-8-7-12-3-2-4-13-9-10-15(11)17(14)16(12)13/h2-10H,1H3 |
| SMILES | Cc1ccc2ccc3cccc4ccc1c2c34 |
| IUPAC Name | 1-methylpyrene |
| Molecular Weight (g/mol) | 216.28 |
| Formula Weight | 216.0939 |