missing translation for 'onlineSavingsMsg'
Learn More

1-Methylhistamine Dihydrochloride, TRC

Product Code. 30138408
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
100mg
10mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30138408 25 mg 25mg
30136871 100 mg 100mg
30005530 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138408 Supplier Toronto Research Chemicals Supplier No. TRCM31215325MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6481-48-7
Molecular Formula C6 H11 N3 . 2HCl
Molecular Weight (g/mol) 198.09352
Synonym 4-(2-Aminoethyl)-1-methyl-imidazole Dihydrochloride, 1-Methyl-1H-imidazole-4-ethanamine Dihydrochloride;
IUPAC Name 2-(1-methylimidazol-4-yl)ethanamine;dihydrochloride
SMILES Cl.Cl.Cn1cnc(CCN)c1

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C6H11N3.2ClH/c1-9-4-6(2-3-7)8-5-9;;/h4-5H,2-3,7H2,1H3;2*1H
Formula Weight 197.0487
Chemical Name or Material 1-Methylhistamine Dihydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.