missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl Chrysene, TRC

Product Code. 18002950
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18002950 10 mg 10mg
17949969 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18002950 Supplier Toronto Research Chemicals Supplier No. TRCM26511510MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3351-28-8
Molecular Formula C19 H14
Molecular Weight (g/mol) 242.31
Synonym Chrysene, 1-methyl-, 1-Methylchrysene
IUPAC Name 1-methylchrysene
SMILES Cc1cccc2c1ccc3c4ccccc4ccc23

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C19H14/c1-13-5-4-8-17-15(13)11-12-18-16-7-3-2-6-14(16)9-10-19(17)18/h2-12H,1H3
Formula Weight 242.1096
Chemical Name or Material 1-Methylchrysene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.