missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl Adamantane, TRC

Product Code. 30471933
Change view
Click to view available options
Quantity:
100 mg
5 g
Unit Size:
100mg
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30471933 100 mg 100mg
30466167 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30471933 Supplier Toronto Research Chemicals Supplier No. TRCM320050100MG
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 768-91-2
Molecular Formula C11 H18
Molecular Weight (g/mol) 150.26
Synonym 1-Methyl-tricyclo[3.3.1.13,7]decane, 1-Methyl-adamantane
IUPAC Name 1-methyladamantane
SMILES C[C@@]12C[C@@H]3C[C@@H](C[C@@H](C3)C1)C2

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C11H18/c1-11-5-8-2-9(6-11)4-10(3-8)7-11/h8-10H,2-7H2,1H3/t8-,9+,10-,11+
Formula Weight 150.1409
Chemical Name or Material 1-Methyl Adamantane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.