missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-3-phenyl-1H-pyrazole-5-carbaldehyde, 97%, Thermo Scientific™

Product Code. 10051534
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10051534 1 g 1g
10411144 250 mg 250mg
10302305 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10051534 Supplier Thermo Scientific Maybridge Supplier No. CC46604DA
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 864068-96-2
Molecular Formula C11H10N2O
Molecular Weight (g/mol) 186.21
MDL Number MFCD08271941
InChI Key XFGHBJQGDDYIKS-UHFFFAOYSA-N
Synonym 1-methyl-3-phenyl-1h-pyrazole-5-carbaldehyde, 1-methyl-3-phenylpyrazole-5-carbaldehyde, 2-methyl-5-phenyl-2h-pyrazole-3-carbaldehyde, 5-formyl-1-methyl-3-phenyl-1h-pyrazole, 1h-pyrazole-5-carboxaldehyde,1-methyl-3-phenyl
PubChem CID 7537629
SMILES CN1N=C(C=C1C=O)C1=CC=CC=C1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
IUPAC Name 2-methyl-5-phenylpyrazole-3-carbaldehyde
Formula Weight 186.21
Percent Purity 97%
Chemical Name or Material 1-Methyl-3-phenyl-1H-pyrazole-5-carbaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.