missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-1H-pyrazole-5-carboxylic acid, 97%, Thermo Scientific™

Product Code. p-7031705
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15435637 250 mg 250mg
15425637 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15435637 Supplier Thermo Scientific Alfa Aesar Supplier No. H32874.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16034-46-1
Molecular Formula C5H6N2O2
Molecular Weight (g/mol) 126.12
MDL Number MFCD00464253
InChI Key JREJQAWGQCMSIY-UHFFFAOYSA-N
Synonym 1-methyl-1h-pyrazole-5-carboxylic acid, 1-methylpyrazole-5-carboxylic acid, 2-methyl-2h-pyrazole-3-carboxylic acid, 1h-pyrazole-5-carboxylic acid, 1-methyl, 1-methyl-1h-pyrazole-5-carboxylicacid, 5-carboxy-1-methyl-1h-pyrazole, 1-methyl-5-pyrazolecarboxylic acid, 2h-pyrazole-3-carboxylic acid, 2-methyl, acmc-209dkx, ksc173g6t
PubChem CID 643158
ChEBI CHEBI:74737
IUPAC Name 2-methylpyrazole-3-carboxylic acid
SMILES CN1N=CC=C1C(O)=O

Specifications

Melting Point 222°C to 225°C
Odor Odorless
Quantity 250 mg
Formula Weight 126.11
Percent Purity 97%
Chemical Name or Material 1-Methyl-1H-pyrazole-5-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.