missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ COMMD6 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15695125
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15695125 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15695125 Supplier Invitrogen™ Supplier No. PA551207
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

The antibody detects endogenous levels of total COMMD6 protein.

COMMD6 belongs to a family of NF-kappa-B (see RELA; MIM 164014)-inhibiting proteins characterized by the presence of a COMM domain (see COMMD1; MIM 607238) (de Bie et al., 2006 [PubMed 16573520]).
TRUSTED_SUSTAINABILITY

Specifications

Antigen COMMD6
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Gene COMMD6
Gene Accession No. Q7Z4G1
Gene Alias Acrg; Acrg embryonic lethality minimal region ortholog; COMM domain containing 6; COMM domain-containing protein 6; COMMD6; MSTP076
Gene Symbols COMMD6
Host Species Rabbit
Immunogen Full length fusion protein of human COMMD6.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 170622
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.