Learn More
1-Methoxy-2-propanol, Supelco™
Product Code. 15878050
Change view
Click to view available options
Quantity:
1 L
25 L
190 L
Unit Size:
1 L
190 L
25 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15878050 190 L 190 L
15858050 1 L 1 L
15868050 25 L 25 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15878050 Supplier Supelco Supplier No. 1167389190
 Restricted Item
Estimated Shipment: 23-11-2026
EMPLURA™
EMPLURA™ is our low-cost alternative to high-purity qualities. With EMPLURA we offer a range of solvents for a plurality of basic applications in non-regulated industries and for less demanding applications, preparative laboratory work and cleaning purposes. EMPLURA solvents provide adequate specifications with the most important parameters.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 107-98-2
Grade Research
Molecular Weight (g/mol) 90.12
Specifications
Melting Point -97°C
Density 0.92 g/cm3 at 20°C
Quantity 190 L
MDL Number MFCD00004537
Synonym 1-Methoxy-2-propanol, Propylene glycol monomethyl ether, 1-Methoxy-2-propanol
Boiling Point 120°C/1013 mbar
Chemical Name or Material 1-Methoxy-2-Propanol