missing translation for 'onlineSavingsMsg'
Learn More

1-Hexanol, ≥99%, MP Biomedicals™

Product Code. 11433842 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11433842 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11433842 Supplier MP Biomedicals Supplier No. 0215736280
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 111-27-3
Color Colorless
Density 0.82 at 20°C
Assay Percent Range ≥99%
Molecular Formula C6H14O
Quantity 100 mL
Synonym 1-hexanol, hexyl alcohol, hexanol, n-hexanol, n-hexyl alcohol, 1-hexyl alcohol, 1-hydroxyhexane, amylcarbinol, caproyl alcohol, pentylcarbinol
InChI Key ZSIAUFGUXNUGDI-UHFFFAOYSA-N
SMILES CCCCCCO
IUPAC Name hexan-1-ol
Molecular Weight (g/mol) 102.177
PubChem CID 8103
ChEBI CHEBI:87393
Formula Weight 102.2
Physical Form Liquid
Chemical Name or Material 1-Hexanol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.