Learn More
1-Hepten-3-ol, 98%
Product Code. 11479110
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11479110 50 g 50g
11469110 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11479110 Supplier Thermo Scientific Alfa Aesar Supplier No. A14651.18
Request Bulk or Custom Format
CAS: 4938-52-7 | C7H14O | 114.188 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 4938-52-7
Molecular Formula C7H14O
Molecular Weight (g/mol) 114.188
MDL Number MFCD00021940
InChI Key PZKFYTOLVRCMOA-UHFFFAOYSA-N
Synonym 1-hepten-3-ol, heptene-1-ol-3, butyl vinyl carbinol, acmc-1aoyf, 1-hepten-3-ol, analytical standard, 1-hepten-3-ol, fg, 1-hepten-3-ol, purum gc
PubChem CID 21057
IUPAC Name hept-1-en-3-ol
SMILES CCCCC(C=C)O
Specifications
Density 0.836
Boiling Point 154°C to 156°C
Flash Point 54°C (129°F)
Refractive Index 1.433
Quantity 50 g
UN Number UN1987
Beilstein 1720340
Formula Weight 114.18
Percent Purity 98%
Chemical Name or Material 1-Hepten-3-ol