Learn More
1-Heptanol, Sigma-Aldrich™
Product Code. 15841081
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
Unit Size:
1 L
100mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15841081 1 L 1 L
15821081 100 mL 100mL
15831081 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15841081 Supplier Sigma Aldrich Supplier No. 8206241000
 Restricted Item
Estimated Shipment: 25-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 111-70-6
Grade Research
Molecular Formula CH3(CH2)6OH
Molecular Weight (g/mol) 116.2
Specifications
Melting Point -35°C
Density 0.82 g/cm3 at 20°C
Boiling Point 176°C/1013 hPa
MDL Number MFCD00002986
Quantity 1 L
Synonym 1-Heptanol, Heptyl alcohol, 1-Heptanol