missing translation for 'onlineSavingsMsg'
Learn More

1-Fluoro-3-nitrobenzene, 98+%

Product Code. p-7028312
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11447236 25 g 25g
11457236 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11447236 Supplier Thermo Scientific Alfa Aesar Supplier No. B25553.14
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 402-67-5 | C6H4FNO2 | 141.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 402-67-5
Molecular Formula C6H4FNO2
Molecular Weight (g/mol) 141.10
MDL Number MFCD00007196
InChI Key WMASLRCNNKMRFP-UHFFFAOYSA-N
Synonym 3-fluoronitrobenzene, m-fluoronitrobenzene, m-nitrofluorobenzene, benzene, 1-fluoro-3-nitro, 3-fluoro-1-nitrobenzene, 1-fluoro-3-nitro-benzene, 3-fluoro-nitrobenzene, 3-nitro-fluorobenzene, pubchem2286, 3-fluoro nitro benzene
PubChem CID 9823
IUPAC Name 1-fluoro-3-nitrobenzene
SMILES [O-][N+](=O)C1=CC=CC(F)=C1

Specifications

Melting Point 2°C
Density 1.325
Boiling Point 204°C to 206°C
Flash Point 76°C (168°F)
Refractive Index 1.525
Quantity 25 g
UN Number UN2810
Beilstein 1862210
Formula Weight 141.1
Percent Purity ≥98%
Chemical Name or Material 1-Fluoro-3-nitrobenzene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.