missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-Ethyl-2-iodobenzene, 98%

Product Code. 11441858
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11441858 250 g 250g
11421858 10 g 10g
11431858 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11441858 Supplier Alfa Aesar™ Supplier No. A14509.30
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Applications
1-Ethyl-2-iodobenzene is used as pharmaceutical intermediate and as a reagent. It is used to prepare ethyl-2′-isopropylbiphenyl.

Solubility
Insoluble in water.

Notes
Light sensitive. Store in the dark. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18282-40-1
Molecular Formula C8H9I
Molecular Weight (g/mol) 232.064
MDL Number MFCD00040871
InChI Key ZEJZDNMOGNUIHL-UHFFFAOYSA-N
PubChem CID 140367
IUPAC Name 1-ethyl-2-iodobenzene
SMILES CCC1=CC=CC=C1I

Specifications

Boiling Point 221°C
Flash Point 101°C (213°F)
Quantity 250 g
Beilstein 2077313
Synonym 2-ethyliodobenzene, 1-iodo-2-ethylbenzene, benzene, 1-ethyl-2-iodo, 2-ethyl iodobenzene, pubchem3876, 1-ethyl-2-iodo-benzene, acmc-1brg2, #, benzene,1-ethyl-2-iodo, attercop-chm at134049
Formula Weight 232.07
Percent Purity 98%
Chemical Name or Material 1-Ethyl-2-iodobenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.