Learn More
1-epi-Ramipril, TRC
Product Code. 30428530
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30428530 100 mg 100mg
30418298 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30428530 Supplier Toronto Research Chemicals Supplier No. TRCR110980100MG
 Restricted Item
Estimated Shipment: 11-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 104195-90-6
Molecular Formula C23H32N2O5
Molecular Weight (g/mol) 416.51
Synonym (2S,3aS,6aS)-1-[(2S)-2-[[(1R)-1-(Ethoxycarbonyl)-3-phenylpropyl]amino]-1-oxopropyl]octahydrocyclopenta[b]pyrrole-2-carboxylic Acid, [2S-[1[R*(S*)],2alpha,3abeta,6abeta]]-1-[2-[[1-(Ethoxycarbonyl)-3-phenylpropyl]amino]-1-oxopropyl]octahydrocyclopenta[b]pyrrole-2-carboxylic Acid, (1R)-epi-Ramipril
IUPAC Name (2S,3aS,6aS)-1-[(2S)-2-[[(2R)-1-ethoxy-1-oxo-4-phenylbutan-2-yl]amino]propanoyl]-3,3a,4,5,6,6a-hexahydro-2H-cyclopenta[b]pyrrole-2-carboxylic acid
SMILES CCOC(=O)[C@@H](CCc1ccccc1)N[C@@H](C)C(=O)N2[C@H]3CCC[C@H]3C[C@H]2C(=O)O
Specifications
Quantity 100 mg
InChI Formula InChI=1S/C23H32N2O5/c1-3-30-23(29)18(13-12-16-8-5-4-6-9-16)24-15(2)21(26)25-19-11-7-10-17(19)14-20(25)22(27)28/h4-6,8-9,15,17-20,24H,3,7,10-14H2,1-2H3,(H,27,28)/t15-,17-,18+,19-,20-/m0/s1
Formula Weight 416.2311
Chemical Name or Material 1-epi-Ramipril