missing translation for 'onlineSavingsMsg'
Learn More

1-Eicosene, MP Biomedicals™

Product Code. 11431291 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11431291 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11431291 Supplier MP Biomedicals Supplier No. 0215790950
  • ONLINE EXCLUSIVE PRICE
    129.00 EUR
    122.85 EUR / 50g
    Save 6.15 EUR
    4% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 9-3-3452
Color Colorless
Assay Percent Range 99.2%
Molecular Formula C9H16
Quantity 50 g
Synonym 1-nonyne, nonyne, unii-760v9gp5a1, 3-acetylnaphthalene, pubchem13060, acmc-1aglr, 1-nonyne 6ci, 7ci, 8ci, 9ci
InChI Key OSSQSXOTMIGBCF-UHFFFAOYSA-N
SMILES CCCCCCCC#C
IUPAC Name non-1-yne
Molecular Weight (g/mol) 124.23
PubChem CID 18937
Formula Weight 280.5
Physical Form Liquid After Melting
Chemical Name or Material 1-Eicosene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.