Learn More
1-Dodecanethiol, Sigma-Aldrich™
Product Code. 15864541
Change view
Click to view available options
Quantity:
250 mL
1 L
10 L
Unit Size:
1 L
10 L
250mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15864541 1 L 1 L
15854541 250 mL 250mL
15841021 10 L 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15864541 Supplier Sigma-Aldrich Supplier No. 8205441000
 Restricted Item
Estimated Shipment: 21-12-2026
For synthesis
1-Dodecanethiol is an alkyl thiol that can be used as an organic source of sulfur. It is used in the production of synthetic rubber, nonionic detergent, plastics, and pharmaceuticals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 112-55-0
Grade Research
Molecular Formula CH3(CH2)11SH
Molecular Weight (g/mol) 202.4
Specifications
Melting Point -9°C to -7°C
Density 0.845 g/cm3 at 20°C
MDL Number MFCD00004885
Quantity 1 L
Synonym 1-Dodecanethiol, Lauryl mercaptan, Dodecyl mercaptan, 1-Dodecanethiol