Learn More
1-Desmethyl-2-methylpropyl Amorolfine, TRC
Specifications
Specifications
| Chemical Name or Material | 1-Desmethyl-2-methylpropyl Amorolfine |
| CAS | 67468-13-7 |
| Molecular Formula | C21H35NO |
| Quantity | 10 mg |
| Synonym | 4-[3-[4-(1,2-Dimethylpropyl)phenyl]-2-methylpropyl]-2,6-dimethyl-morpholine, Amorolfine EP Impurity I |
| InChI Formula | InChI=1S/C21H35NO/c1-15(2)19(6)21-9-7-20(8-10-21)11-16(3)12-22-13-17(4)23-18(5)14-22/h7-10,15-19H,11-14H2,1-6H3/t16?,17-,18+,19? |
| SMILES | C[C@@H]1CN(CC(C)CC2=CC=C(C(C)C(C)C)C=C2)C[C@H](C)O1 |
| Molecular Weight (g/mol) | 317.51 |