Learn More
1-Decene, Dr. Ehrenstorfer
Product Code. 30309923
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30309923 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30309923 Supplier Dr. Ehrenstorfer Supplier No. DRECA12096000
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 1-Decene
CAS 872-05-9
Analyte or Component Names 1-Decene
Shipping Condition +20°C
Molecular Formula C10 H20
Certifications/Compliance ISO 17034
Synonym 1-Decene, 1-n-Decene, Dialen 10, Gulftene 10, Linealene 10, NSC 62122, Neodene 10, n-1-Decene, α-Decene
InChI Formula InChI=1S/C10H20/c1-3-5-7-9-10-8-6-4-2/h3H,1,4-10H2,2H3
SMILES CCCCCCCCC=C
IUPAC Name dec-1-ene
Molecular Weight (g/mol) 140.27
Quantity 1 mL
Formula Weight 140.1565 g/mol
Show More Show Less
Product Title
Vælg et problem
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.