missing translation for 'onlineSavingsMsg'
Learn More

1-Cyclopropylethylamine Hydrochloride, TRC

Product Code. 30362765
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
Unit Size:
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30362765 250 mg 250mg
30360113 500 mg -
30348906 100 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30362765 Supplier Toronto Research Chemicals Supplier No. TRCC986725250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 42390-64-7
Molecular Formula C5H11N . HCl
Molecular Weight (g/mol) 121.61
Synonym Cyclopropanemethanamine, α-methyl-, hydrochloride (1:1) (ACI), Cyclopropanemethanamine, α-methyl-, hydrochloride (9CI), 1-Cyclopropylethan-1-amine hydrochloride, 1-Cyclopropylethylamine hydrochloride, α-Methylcyclopropanemethanamine hydrochloride
IUPAC Name 1-cyclopropylethanamine;hydrochloride
SMILES Cl.CC(N)C1CC1

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C5H11N.ClH/c1-4(6)5-2-3-5;/h4-5H,2-3,6H2,1H3;1H
Formula Weight 121.0658
Chemical Name or Material 1-Cyclopropylethylamine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.