Learn More
1-Chlorobutane, Supelco™
Description
With their high degree of UV transmittance, low particle count, low acidity and alkalinity and low evaporation residue level, LiChrosolv solvents are ideal for reproducible separations. Since separations are normally carried out under gradient conditions in analytical HPLC, we offer solvents in \gradient grade\ as well as \isocratic grade\. This enables to minimize the gradient effect of the solvent involved.
Specifications
Specifications
| CAS | 109-69-3 |
| Melting Point | -123°C |
| Density | 0.886 g/cm3 at 20°C |
| Boiling Point | 79°C/1013 hPa |
| Molecular Formula | CH3(CH2)3Cl |
| MDL Number | MFCD00001009 |
| Quantity | 1 L |
| Synonym | 1-Chlorobutane, n-Butyl chloride, 1-Chlorobutane |
| Molecular Weight (g/mol) | 92.57 |
| Grade | Research |