missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Chlorobutane, For Synthesis, MilliporeSigma™
Specifications
Specifications
| CAS | 109-69-3 |
| Melting Point | -123°C |
| Packaging | Glass Bottle |
| Molecular Formula | C4H9Cl |
| MDL Number | MFCD00001009 |
| Quantity | 1 L |
| UN Number | 1127 |
| Synonym | butyl chloride, n-butyl chloride, butane, 1-chloro, chlorure de butyle, nbc wormer, n-propylcarbinyl chloride, sure shot, butane, chloro, chlorobutane, n-chlorobutane |
| InChI Key | VFWCMGCRMGJXDK-UHFFFAOYSA-N |
| SMILES | CCCCCl |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?