missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-3,5-dimethyladamantane, TRC

Product Code. 30012548
Change view
Click to view available options
Quantity:
50 mg
250 mg
1 g
Unit Size:
1g
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30012548 1 g 1g
30006220 50 mg 50mg
30006557 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30012548 Supplier Toronto Research Chemicals Supplier No. TRCC3653801G
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 707-36-8
Molecular Formula C12 H19 Cl
Molecular Weight (g/mol) 198.73
Synonym 1-Chloro-3,5-dimethyladamantane, Memantine Related Compound C (USP)
IUPAC Name 1-chloro-3,5-dimethyladamantane
SMILES CC12CC3CC(C)(C1)CC(Cl)(C3)C2

Specifications

Quantity 1 g
InChI Formula InChI=1S/C12H19Cl/c1-10-3-9-4-11(2,6-10)8-12(13,5-9)7-10/h9H,3-8H2,1-2H3
Formula Weight 198.1175
Chemical Name or Material 1-Chloro-3,5-dimethyladamantane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.