Learn More
Thermo Scientific™ Cartridges for Multidrop™ Pico Digital Dispensers
Alle ansehen Thermo Scientific Produkte
Beschreibung
The cartridges are designed for use with Multidrop™ Pico Digital Dispensers, dispensing volumes from 8 pL to 10 μL. Use the 1-channel cartridges with the Multidrop™ Pico 1, and the 4- or 8-channel standard or surfactant free cartridges with the Multidrop™ Pico 8 Surfactant Free (Pico 8 sf).
Features:
- Single-Channel Cartridges: Use the 1-channel cartridges with the Multidrop™ Pico 1 for single fluid dispensing at a time.
- Multi-Channel Cartridges: Use the 4- or 8-channel cartridges with the Multidrop™ Pico 8 sf to dispense multiple fluids per cartridge.
- Surfactant Free Cartridges: The Multidrop™ Pico 8 sf with the surfactant free cartridges can be used to dispense aqueous fluids without surfactant.
- Contamination-Free Dispensing: Non-contact dispensing and single-use cartridges prevent carry-over and contamination.
- Fluid Compatibility: Ideal for DMSO-based fluids, aqueous fluids, or master mixes, with or without small molecules or biomolecules. Aqueous fluids can be dispensed without surfactant when using the surfactant free cartridges with the Multidrop™ Pico 8 sf.
Spezifikation
Spezifikation
| Product Type | Digital Dispenser Cartridge |
| Channels | 4 |
| Volume (Metric) | 1 nL to 10 μL |
| For Use With (Equipment) | Multidrop Pico 8 Digital Dispenser, Multidrop Pico 8 Surfactant Free Digital Dispenser |
| Model | 4-channel 200 μL Standard Cartridges |
| No. per Pack | Case of 20 |
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.