Learn More
(1-Butyl)triethylammonium bromide, 97%
Product Code. p-7027404
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11461137 25 g 25g
11471137 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11461137 Supplier Thermo Scientific Alfa Aesar Supplier No. B24128.14
Add to basket
CAS: 13028-69-8 | C10H24BrN | 238.213 g/mol
(1-Butyl)triethylammonium bromide is an important organic intermediate. It can be used in agrochemical, pharmaceutical & dye stuff field.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
(1-Butyl)triethylammonium bromide is an important organic intermediate. It can be used in agrochemical, pharmaceutical & dye stuff field.
Solubility
Soluble in water.
Notes
Hygroscopic. Store away from oxidizing agents, water/ moisture. Store under inert gas.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 13028-69-8
Molecular Formula C10H24BrN
Molecular Weight (g/mol) 238.213
MDL Number MFCD00055541
InChI Key ZNFYPPCUDSCDPR-UHFFFAOYSA-M
Synonym 1-butyl triethylammonium bromide, butyltriethylammonium bromide, n,n,n-triethylbutan-1-aminium bromide, butyltriethylazanium bromide, acmc-20apri, triethylbutyl ammonium bromide, butyl-triethyl-ammonium bromide, triethyl-n-butylammonium bromide, 1-butanaminium,n,n,n-triethyl-, bromide 1:1
PubChem CID 11746670
IUPAC Name butyl(triethyl)azanium;bromide
SMILES CCCC[N+](CC)(CC)CC.[Br-]
Specifications
Melting Point 212°C to 215°C
Quantity 25 g
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 238.21
Percent Purity 97%
Chemical Name or Material (1-Butyl)triethylammonium bromide