missing translation for 'onlineSavingsMsg'
Learn More

1-Butyl-2,3-dimethylimidazolium hexafluorophosphate, 99%

Product Code. 11387399
Click to view available options
Quantity:
5 g
50 g
Unit Size:
50g
5g
This item is not returnable. View return policy

Product Code. 11387399

Brand: Thermo Scientific Alfa Aesar H27827.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 227617-70-1 | C9H17F6N2P | 298.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 227617-70-1
Molecular Formula C9H17F6N2P
Molecular Weight (g/mol) 298.21
MDL Number MFCD03790877
InChI Key JWFPQAXAGSAKRF-UHFFFAOYSA-N
Synonym 1-butyl-2,3-dimethylimidazolium hexafluorophosphate, 1-butyl-2,3-dimethyl-1h-imidazol-3-ium hexafluorophosphate v, 1-butyl-2,3-dimethylimidazol-1-ium hexafluorophosphate, 3-butyl-1,2-dimethylimidazol-1-ium hexafluorophosphate, acmc-209fyn, bdmim pf6, c4c1mim pf6, c4mmim pf6, dsstox_cid_27883, dsstox_rid_82636
PubChem CID 11243457
IUPAC Name 1-butyl-2,3-dimethylimidazol-3-ium;hexafluorophosphate
SMILES F[P-](F)(F)(F)(F)F.CCCCN1C=C[N+](C)=C1C

Specifications

Melting Point 48°C to 50°C
Density 1.354
Refractive Index 1.423
Quantity 5 g
Formula Weight 298.21
Percent Purity 99%
Chemical Name or Material 1-Butyl-2,3-dimethylimidazolium hexafluorophosphate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.