CAS RN 227617-70-1
CAS RN 227617-70-1
IUPAC Name:
1-butyl-2,3-dimethyl-1H-imidazol-3-ium; hexafluoro-λ⁵-phosphanuide
Synonyms:
3-butyl-1,2-dimethylimidazol-1-ium hexafluorophosphate
Molecular Weight (g/mol):
298.21
Molecular Formula:
C9H17F6N2P
InChi Key:
JWFPQAXAGSAKRF-UHFFFAOYSA-N
SMILES:
F[P-](F)(F)(F)(F)F.CCCCN1C=C[N+](C)=C1C
1
–
1
of
1
results
1
1-Butyl-2,3-dimethylimidazolium hexafluorophosphate, 99%
CAS: 227617-70-1 Molecular Formula: C9H17F6N2P Molecular Weight (g/mol): 298.21 MDL Number: MFCD03790877 InChI Key: JWFPQAXAGSAKRF-UHFFFAOYSA-N Synonym: 1-butyl-2,3-dimethylimidazolium hexafluorophosphate,1-butyl-2,3-dimethyl-1h-imidazol-3-ium hexafluorophosphate v,1-butyl-2,3-dimethylimidazol-1-ium hexafluorophosphate,3-butyl-1,2-dimethylimidazol-1-ium hexafluorophosphate,acmc-209fyn,bdmim pf6,c4c1mim pf6,c4mmim pf6,dsstox_cid_27883,dsstox_rid_82636 PubChem CID: 11243457 IUPAC Name: 1-butyl-2,3-dimethylimidazol-3-ium;hexafluorophosphate SMILES: F[P-](F)(F)(F)(F)F.CCCCN1C=C[N+](C)=C1C