missing translation for 'onlineSavingsMsg'
Learn More

1-Bromoundecane, 98%

Product Code. p-7021836
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11426897 25 g 25g
11436897 100 g 100g
11446897 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11426897 Supplier Thermo Scientific Alfa Aesar Supplier No. A14333.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 693-67-4 | C11H23Br | 235.209 g/mol

1-Bromoundecane is used in the preparation of Grignard reagent by reacting with Mg in THF (tetrahydrofuran). It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, dyes and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromoundecane is used in the preparation of Grignard reagent by reacting with Mg in THF (tetrahydrofuran). It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, dyes and agrochemicals.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 693-67-4
Molecular Formula C11H23Br
Molecular Weight (g/mol) 235.209
MDL Number MFCD00000223
InChI Key IKPSIIAXIDAQLG-UHFFFAOYSA-N
Synonym undecyl bromide, undecane, 1-bromo, hendecyl bromide, n-undecyl bromide, bromoundecane, 1-bromo-undecane, n-undecyl-1-bromide, labotest-bb ltbb001165, undecylbromide, 1-bromanylundecane
PubChem CID 12744
IUPAC Name 1-bromoundecane
SMILES CCCCCCCCCCCBr

Specifications

Melting Point -9°C
Density 1.052
Boiling Point 112°C to 114°C (10 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.4563
Quantity 25 g
Beilstein 1098390
Solubility Information Insoluble in water.
Formula Weight 235.22
Percent Purity 98%
Chemical Name or Material 1-Bromoundecane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.