accessibility menu, dialog, popup

UserName

CAS RN 693-67-4

H3CBr

CAS RN 693-67-4

IUPAC Name: 1-bromoundecane
Synonyms: 1-bromoundecane
Molecular Weight (g/mol): 235.21
Molecular Formula: C11H23Br
InChi Key: IKPSIIAXIDAQLG-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCBr
Molecular Weight (g/mol) 
  • (4)
  • (1)
Percent Purity 
  • (1)
  • (4)
Quantity 
  • (2)
  • (2)
  • (1)
Boiling Point 
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (4)
  • (1)
  • (1)
  • (4)
  • (2)
  • (2)
  • (1)
  • (3)
  • (1)
  • (2)