Learn More
1-Bromonaphthalene, Sigma-Aldrich™
Product Code. 15894181
Change view
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
Unit Size:
100mL
2.5 L
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15894181 100 mL 100mL
15804191 500 mL 500mL
15814191 2.5 L 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15894181 Supplier Sigma Aldrich Supplier No. 8062100100
 Restricted Item
Estimated Shipment: 18-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 90-11-9
Grade Research
Molecular Weight (g/mol) 207.07
Specifications
Density 1.48 g/cm3 at 20°C
Boiling Point 281°C/1013 hPa
MDL Number MFCD00003868
Quantity 100 mL
Synonym 1-Bromonaphthalene, α-Naphthyl bromide, 1-Bromonaphthalene