missing translation for 'onlineSavingsMsg'
Learn More

1-Bromohexane, 99%

Product Code. 11492158
Change view
Click to view available options
Quantity:
10,000 g
100 g
2500 g
500 g
Unit Size:
10000g
100g
2500g
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11492158 500 g 500g
11474180 100 g 100g
11402168 2500 g 2500g
11484180 10,000 g 10000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11492158 Supplier Thermo Scientific Alfa Aesar Supplier No. A15166.36
  • ONLINE EXCLUSIVE PRICE
    54.10 EUR
    48.65 EUR / 500g
    Save 5.45 EUR
    10% Off
 Restricted Item
Estimated Shipment: 28-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 111-25-1 | C6H13Br | 165.07 g/mol

1-Bromohexane is utilized in the preparation of pharmaceuticals and organic chemicals. It is involved in Grignard reactions as an alkylating agent and in the synthesis of bromo-cyclic compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromohexane is utilized in the preparation of pharmaceuticals and organic chemicals. It is involved in Grignard reactions as an alkylating agent and in the synthesis of bromo-cyclic compounds.

Solubility
Immiscible with water.

Notes
Keep away from heat and sources of ignition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-25-1
Molecular Formula C6H13Br
Molecular Weight (g/mol) 165.07
MDL Number MFCD00000271
InChI Key MNDIARAMWBIKFW-UHFFFAOYSA-N
Synonym hexyl bromide, n-hexyl bromide, bromohexane, 1-hexyl bromide, hexane, 1-bromo, hexane, bromo, unii-wva0fax7ga, 1-bromo-hexane, pentane, bromomethyl, wva0fax7ga
PubChem CID 8101
IUPAC Name 1-bromohexane
SMILES CCCCCCBr

Specifications

Melting Point -85°C
Density 1.176
Boiling Point 156°C
Flash Point 50°C (122°F)
Odor Pleasant
Refractive Index 1.4475
Quantity 500 g
UN Number UN1993
Beilstein 1731290
Solubility Information Immiscible with water.
Formula Weight 165.08
Percent Purity 99%
Chemical Name or Material 1-Bromohexane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.