missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-4-methylcyclohexane, cis + trans, 97%

Product Code. p-7043772
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11307785 5 g 5g
11317785 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11307785 Supplier Thermo Scientific Alfa Aesar Supplier No. L02640.06
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 6294-40-2 | C7H13Br | 177.085 g/mol

1-Bromo-4-methylcyclohexane is used as intermediates in pharmaceutical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromo-4-methylcyclohexane is used as intermediates in pharmaceutical research.

Solubility
Not miscible in water.

Notes
Store away from oxidizing agents. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6294-40-2
Molecular Formula C7H13Br
Molecular Weight (g/mol) 177.085
MDL Number MFCD00014282
InChI Key RLOHLPHAOHGRNM-UHFFFAOYSA-N
Synonym cyclohexane, 1-bromo-4-methyl, p-methylcyclohexyl bromide, 4-bromo-1-methylcyclohexane, 4-methylcyclohexyl bromide, acmc-1b9h5, 1-bromo-4-methyl-cyclohexane, 4-methylcyclohexyl bromide cis+trans, cyclohexane, 1-bromo-4-methyl-, trans, 1-bromo-4-methylcyclohexane, cis + trans
PubChem CID 22727
IUPAC Name 1-bromo-4-methylcyclohexane
SMILES CC1CCC(CC1)Br

Specifications

Density 1.262
Boiling Point 94°C to 96°C (50 mmHg)
Refractive Index 1.488
Quantity 5 g
Beilstein 1900954
Solubility Information Not miscible in water.
Formula Weight 177.09
Percent Purity 97%
Chemical Name or Material 1-Bromo-4-methylcyclohexane, Cis + Trans

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.