missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-4-difluoromethyl-2-iodobenzene, 97%, Thermo Scientific™

Product Code. 15466177
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Product Code. Quantity unitSize
15466177 250 mg 250mg
15456177 1 g 1g
2 options
This item is not returnable. View return policy

Product Code. 15466177

Brand: Thermo Scientific Alfa Aesar H33360.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1261671-65-1
Molecular Formula C7H4BrF2I
Molecular Weight (g/mol) 332.914
MDL Number MFCD18391107
InChI Key IKWVZHWJWGKEFJ-UHFFFAOYSA-N
Synonym 1-bromo-4-difluoromethyl-2-iodobenzene
PubChem CID 86276462
IUPAC Name 1-bromo-4-(difluoromethyl)-2-iodobenzene
SMILES C1=CC(=C(C=C1C(F)F)I)Br

Specifications

Refractive Index 1.603
Quantity 250 mg
Sensitivity Light sensitive
Formula Weight 332.91
Percent Purity 97%
Chemical Name or Material 1-Bromo-4-difluoromethyl-2-iodobenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.