accessibility menu, dialog, popup

UserName

CAS RN 1261671-65-1

FFIBr

CAS RN 1261671-65-1

IUPAC Name: 1-bromo-4-(difluoromethyl)-2-iodobenzene
Synonyms: 1-bromo-4-(difluoromethyl)-2-iodobenzene
Molecular Weight (g/mol): 332.91
Molecular Formula: C7H4BrF2I
InChi Key: IKWVZHWJWGKEFJ-UHFFFAOYSA-N
SMILES: FC(F)C1=CC(I)=C(Br)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results