missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-3-(trifluoromethoxy)benzene, 98+%, Thermo Scientific Chemicals

Product Code. 11319519
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11319519 1 g 1g
11407887 5 g 5g
11417887 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11319519 Supplier Thermo Scientific Alfa Aesar Supplier No. A16561.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2252-44-0 | Molecular Formula: C7H4BrF3O | Molecular Weight: 241.007 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2252-44-0
Molecular Formula C7H4BrF3O
Molecular Weight (g/mol) 241.007
MDL Number MFCD00040943
InChI Key WVUDHWBCPSXAFN-UHFFFAOYSA-N
Synonym 1-bromo-3-trifluoromethoxy benzene, 3-trifluoromethoxy bromobenzene, 3-bromo trifluoromethoxy benzene, m-bromo trifluoromethoxy benzene, 3-bromo-1-trifluoromethoxy benzene, benzene, 1-bromo-3-trifluoromethoxy, 3-bromo-alpha,alpha,alpha-trifluoroanisole, bromo-3-trifluoromethoxy benzene, buttpark 91\57-27, 1-bromo-3-trifluoromethoxy-benzene
PubChem CID 519964
IUPAC Name 1-bromo-3-(trifluoromethoxy)benzene
SMILES C1=CC(=CC(=C1)Br)OC(F)(F)F

Specifications

Density 1.62
Boiling Point 156°C to 158°C
Flash Point 62°C (143°F)
Refractive Index 1.462
Quantity 1 g
Beilstein 1945938
Formula Weight 241.01
Percent Purity ≥98%
Chemical Name or Material 1-Bromo-3-(trifluoromethoxy)benzene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.