Learn More
1-Bromo-3,5-dimethyladamantane, TRC
Product Code. 30137356
Change view
Click to view available options
Quantity:
50 mg
1 g
5 g
Unit Size:
1g
50mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137356 5 g 5g
17940015 50 mg 50mg
17948804 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30137356 Supplier Toronto Research Chemicals Supplier No. TRCB6841205G
 Restricted Item
Estimated Shipment: 10-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 941-37-7
Molecular Formula C12 H19 Br
Molecular Weight (g/mol) 243.18
Synonym 1-Bromo-3,5-dimethyladamantane, Memantine Related Compound D (USP)
IUPAC Name 1-bromo-3,5-dimethyladamantane
SMILES CC12CC3CC(C)(C1)CC(Br)(C3)C2
Specifications
Quantity 5 g
InChI Formula InChI=1S/C12H19Br/c1-10-3-9-4-11(2,6-10)8-12(13,5-9)7-10/h9H,3-8H2,1-2H3
Formula Weight 242.067
Percent Purity >95%
Chemical Name or Material 1-Bromo-3,5-dimethyladamantane
Grade GC