Learn More
1-Bromo-2,3-dichlorobenzene, 98%
Product Code. 11464977
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11464977 25 g 25g
11474977 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11464977 Supplier Thermo Scientific Alfa Aesar Supplier No. B21904.14
 Restricted Item
Estimated Shipment: 07-09-2026
Add to basket
CAS: 56961-77-4 | C6H3BrCl2 | 225.89 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 56961-77-4
Molecular Formula C6H3BrCl2
Molecular Weight (g/mol) 225.89
MDL Number MFCD00000536
InChI Key HVKCZUVMQPUWSX-UHFFFAOYSA-N
Synonym 2,3-dichlorobromobenzene, benzene, 1-bromo-2,3-dichloro, benzene, bromodichloro, 2,3-dichloro-1-bromobenzene, 1-bromo-2,3-dichloro-benzene, attercop-chm at113305, bromodichlorobenzene, pubchem3607, monobromodichlorobenzene, acmc-209lv2
PubChem CID 42066
IUPAC Name 1-bromo-2,3-dichlorobenzene
SMILES ClC1=CC=CC(Br)=C1Cl
Specifications
Melting Point 56°C to 60°C
Boiling Point 243°C
Quantity 25 g
Beilstein 2246806
Formula Weight 225.91
Percent Purity 98%
Chemical Name or Material 1-Bromo-2,3-dichlorobenzene