missing translation for 'onlineSavingsMsg'
Learn More

1-Boc-azetidine-3-carboxylic acid, 97%

Product Code. p-7030455
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15413837 250 mg 250mg
11472160 1 g 1g
11482160 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15413837 Supplier Thermo Scientific Alfa Aesar Supplier No. H28817.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 142253-55-2 | C9H14NO4 | 200.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 142253-55-2
Molecular Formula C9H14NO4
Molecular Weight (g/mol) 200.22
MDL Number MFCD01860897
InChI Key NCADHSLPNSTDMJ-UHFFFAOYSA-M
Synonym 1-boc-azetidine-3-carboxylic acid, 1-n-boc-3-azetidinecarboxylic acid, n-boc-azetidine-3-carboxylic acid, boc-azetidine-3-carboxylic acid, 1-tert-butoxycarbonyl azetidine-3-carboxylic acid, 1-tert-butoxy carbonyl azetidine-3-carboxylic acid, 1-n-boc-azetidine-3-carboxylic acid, boc-aze 3-oh, azetidine-1,3-dicarboxylic acid mono-tert-butyl ester, n-boc-3-azetidine carboxylic acid
PubChem CID 2755981
SMILES CC(C)(C)OC(=O)N1CC(C1)C([O-])=O

Specifications

Melting Point 98°C to 105°C
IUPAC Name 1-[(2-methylpropan-2-yl)oxycarbonyl]azetidine-3-carboxylic acid
Quantity 250 mg
Formula Weight 201.22
Percent Purity 97%
Chemical Name or Material 1-Boc-azetidine-3-carboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.