missing translation for 'onlineSavingsMsg'
Learn More

1-Boc-5-(tert-butyldimethylsiloxy)indole-2-boronic acid, 98%, Thermo Scientific™

Product Code. 15439865
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15439865 1 g 1g
15449865 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15439865 Supplier Thermo Scientific Alfa Aesar Supplier No. H52511.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 335649-61-1
Molecular Formula C19H30BNO5Si
Molecular Weight (g/mol) 391.346
MDL Number MFCD08689532
InChI Key NXKIUYOUKLIWLP-UHFFFAOYSA-N
Synonym 5-tert-butyldimethylsilyloxy-1h-indole-2-boronic acid, n-boc protected, 1-boc-5-tbdmso-indole-2-boronic acid, 1-tert-butoxycarbonyl-5-tert-butyldimethylsilyloxy-1h-indol-2-boronic acid, 1-tert-butoxycarbonyl-5-tert-butyldimethylsilyloxy-1h-indol-2-ylboronic acid, 1-tert-butoxycarbonyl-5-tert-butyl dimethyl silyl oxy-1h-indol-2-yl boronic acid, 1-tert-butoxycarbonyl-5-tert-butyldimethylsilyl oxy indol-2-ylboronic acid, 1-boc-5-tbdmso-2-indoleboronic acid, 1-boc-5-tert-butyldimethylsiloxy indole-2-boronic acid, 1-tert-butoxycarbonyl-5-tert-butyldimethylsilyloxy indole, 5-tert-butyldimethylsilyloxy-1h-indole-2-boronic acid,n-boc protected 98
PubChem CID 22285134
IUPAC Name [5-[tert-butyl(dimethyl)silyl]oxy-1-[(2-methylpropan-2-yl)oxycarbonyl]indol-2-yl]boronic acid
SMILES B(C1=CC2=C(N1C(=O)OC(C)(C)C)C=CC(=C2)O[Si](C)(C)C(C)(C)C)(O)O

Specifications

Melting Point 104°C to 106°C
Quantity 1 g
Formula Weight 391.35
Percent Purity 98%
Chemical Name or Material 1-Boc-5-(tert-butyldimethylsiloxy)indole-2-boronic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.