missing translation for 'onlineSavingsMsg'
Learn More

1-Boc-3-hydroxy-1,2,3,6-tetrahydropyridine, 97%, Thermo Scientific Chemicals

Product Code. p-7034877
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15449915 250 mg 250mg
15429337 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15449915 Supplier Thermo Scientific Alfa Aesar Supplier No. H52577.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 224779-27-5 | Molecular Formula: C10H17NO3 | Molecular Weight: 199.25 g/mol

1-Boc-3-hydroxy-1,2,3,6-tetrahydropyridine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Boc-3-hydroxy-1,2,3,6-tetrahydropyridine is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 224779-27-5
Molecular Formula C10H17NO3
Molecular Weight (g/mol) 199.25
MDL Number MFCD04972246
InChI Key NICJOYHYEDFTIX-UHFFFAOYSA-N
Synonym n-boc-3-hydroxy-1,2,3,6-tetrahydropyridine, tert-butyl 5-hydroxy-5,6-dihydropyridine-1 2h-carboxylate, 1-boc-3-hydroxy-1,2,3,6-tetrahydropyridine, 3-hydroxy-3,6-dihydro-2h-pyridine-1-carboxylic acid tert-butyl ester, 3-hydroxy-1,2,3,6-tetrahydropyridine, n-boc protected, tert-butyl 5,6-dihydro-5-hydroxypyridine-1 2h-carboxylate, tert-butyl 3-hydroxy-3,6-dihydropyridine-1 2h-carboxylate, tert-butyl 3-hydroxy-1,2,3,6-tetrahydropyridine-1-carboxylate, 1 2h-pyridinecarboxylic acid, 3,6-dihydro-3-hydroxy-, 1,1-dimethylethyl ester, 1-boc-3-hydroxy-1,2,3,6-tetrahydro-pyridine
PubChem CID 17750131
IUPAC Name tert-butyl 3-hydroxy-3,6-dihydro-2H-pyridine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CC=CC(C1)O

Specifications

Quantity 250 mg
Formula Weight 199.25
Percent Purity 97%
Chemical Name or Material 1-Boc-3-hydroxy-1,2,3,6-tetrahydropyridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.