Learn More
1-Aminocyclobutanecarboxylic acid, 95%
Product Code. 15431679
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15431679 5 g 5g
15421679 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15431679 Supplier Thermo Scientific Alfa Aesar Supplier No. H64459.06
Request Bulk or Custom Format
CAS: 22264-50-2 | C5H9NO2 | 115.132 g/mol
ACBC is an NMDA receptor antagonist acting at the glycine site. It acts as an agonist being an analog of glycine at the NMDA-glycine receptor site. This affects the signal transmission in the CNS.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
ACBC is an NMDA receptor antagonist acting at the glycine site. It acts as an agonist being an analog of glycine at the NMDA-glycine receptor site. This affects the signal transmission in the CNS.
Solubility
Soluble to 100 mM in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 22264-50-2
Molecular Formula C5H9NO2
Molecular Weight (g/mol) 115.132
MDL Number MFCD00661068
InChI Key FVTVMQPGKVHSEY-UHFFFAOYSA-N
Synonym 1-aminocyclobutanecarboxylic acid, acbc, 1-amino-cyclobutanecarboxylic acid, 1-aminocyclobutane carboxylic acid, 1-amino-1-cyclobutanecarboxylic acid, cyclobutanecarboxylic acid, 1-amino, 1-aminocyclobutanecarboxlic acid, 1-amino-1-carboxycyclobutane, cyclovaline
PubChem CID 89643
IUPAC Name 1-aminocyclobutane-1-carboxylic acid
SMILES C1CC(C1)(C(=O)O)N
Specifications
Melting Point 261°C (decomposition)
Color White
Quantity 5 g
Beilstein 2802253
Solubility Information Soluble to 100 mM in water.
Formula Weight 115.13
Percent Purity 95%
Physical Form Powder
Chemical Name or Material 1-Aminocyclobutanecarboxylic acid
compliance-icons
Product Identifier
  • 1-Aminocyclobutanecarboxylic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 1-Amino-1-cyclobutanecarboxylic acid