CAS RN 22264-50-2
CAS RN 22264-50-2
IUPAC Name:
1-aminocyclobutane-1-carboxylic acid
Synonyms:
1-aminocyclobutane-1-carboxylic acid
Molecular Weight (g/mol):
115.13
Molecular Formula:
C5H9NO2
InChi Key:
FVTVMQPGKVHSEY-UHFFFAOYSA-N
SMILES:
NC1(CCC1)C(O)=O
1
–
1
of
1
results
1
1-Aminocyclobutanecarboxylic acid, 95%
CAS: 22264-50-2 Molecular Formula: C5H9NO2 Molecular Weight (g/mol): 115.132 MDL Number: MFCD00661068 InChI Key: FVTVMQPGKVHSEY-UHFFFAOYSA-N Synonym: 1-aminocyclobutanecarboxylic acid,acbc,1-amino-cyclobutanecarboxylic acid,1-aminocyclobutane carboxylic acid,1-amino-1-cyclobutanecarboxylic acid,cyclobutanecarboxylic acid, 1-amino,1-aminocyclobutanecarboxlic acid,1-amino-1-carboxycyclobutane,cyclovaline PubChem CID: 89643 IUPAC Name: 1-aminocyclobutane-1-carboxylic acid SMILES: C1CC(C1)(C(=O)O)N