Learn More
1-Aminoanthraquinone, Sigma-Aldrich™
Product Code. 15812231
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15812231 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
MQ200 Graded - For Research and Non-regulated Industrial Applications
Product Code. 15812231 Supplier Sigma Aldrich Supplier No. 8415900100
 Restricted Item
Estimated Shipment: 26-11-2026
MQ200 Graded - For Research and Non-regulated Industrial Applications
For synthesis
TRUSTED_SUSTAINABILITY
Specifications
CAS 82-45-1
Melting Point 251°C to 254°C
MDL Number MFCD00001213
Quantity 100 g
Synonym 1-Aminoanthraquinone, 1-Aminoanthraquinone
Molecular Weight (g/mol) 223.23
Grade Research