Learn More
1-Amino-4-methylpiperazine (>90%), TRC
Click to view available options
Quantity:
100 g
Unit Size:
100g
Specifications
Specifications
| Chemical Name or Material | 1-Amino-4-methylpiperazine |
| CAS | 6928-85-4 |
| Molecular Formula | C5H13N3 |
| Quantity | 100 g |
| Synonym | 4-Methyl-1-piperazinamine, (4-Methylpiperazin-1-yl)amine, 1-Methyl-4-aminopiperazine, 4-Amino-1-methylpiperazine, 4-Methyl-1-aminopiperazine, N-Amino-N'-methylpiperazineN1-Methyl-N4-aminopiperazine, NSC 80649 |
| InChI Formula | InChI=1S/C5H13N3/c1-7-2-4-8(6)5-3-7/h2-6H2,1H3 |
| SMILES | CN1CCN(N)CC1 |
| IUPAC Name | 4-methylpiperazin-1-amine |
| Molecular Weight (g/mol) | 115.18 |
| Formula Weight | 115.1109 |