Learn More
1-Allylimidazole, 97%
Product Code. 11387516
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
Product Code. 11387516
Brand: Thermo Scientific Alfa Aesar L11053.09
Request Bulk or Custom Format
CAS: 31410-01-2 | C6H8N2 | 108.14 g/mol
1-Allylimidazole, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Allylimidazole, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
It is soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 31410-01-2
Molecular Formula C6H8N2
Molecular Weight (g/mol) 108.14
MDL Number MFCD00037894
InChI Key XLXCHZCQTCBUOX-UHFFFAOYSA-N
Synonym 1-allylimidazole, 1-allyl-1h-imidazole, 1-prop-2-en-1-yl-1h-imidazole, 1h-imidazole, 1-2-propenyl, 1-prop-2-en-1-yl imidazole, imidazole, 1-allyl, n-allylimidazole, 1-allyl imidazole, acmc-1cr2q, 1-allyl-1h-imidazole #
PubChem CID 35794
IUPAC Name 1-prop-2-enylimidazole
SMILES C=CCN1C=CN=C1
Specifications
Boiling Point 212°C to 213°C
Flash Point 101°C (213°F)
Quantity 10 g
UN Number UN3267
Beilstein 605953
Formula Weight 108.14
Percent Purity 99%
Chemical Name or Material 1-Allylimidazole