Learn More
1-Allyl-3-methylimidazolium chloride, 98%
Product Code. 11461550
Change view
Click to view available options
Quantity:
5 g
50 g
Unit Size:
50g
5g
Product Code. Quantity unitSize
11461550 5 g 5g
11471550 50 g 50g
2 options
Product Code. 11461550
Brand: Thermo Scientific Alfa Aesar H26952.06
Request Bulk or Custom Format
CAS: 65039-10-3 | C7H11ClN2 | 158.629 g/mol
1-Allyl-3-methylimidazolium chloride is used as an ionic liquid as well as nonderivatizing solvent for cellulose. It acts as a plasticizer for cornstarch and finds application as a solid biopolymer electrolyte. Further, it plays an important role in the preparation of reducing sugar.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Allyl-3-methylimidazolium chloride is used as an ionic liquid as well as nonderivatizing solvent for cellulose. It acts as a plasticizer for cornstarch and finds application as a solid biopolymer electrolyte. Further, it plays an important role in the preparation of reducing sugar.
Notes
Moisture sensitive. Store in a cool place. Incompatible with strong oxidizing agents and bases.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 65039-10-3
Molecular Formula C7H11ClN2
Molecular Weight (g/mol) 158.629
MDL Number MFCD07784450
InChI Key QVRCRKLLQYOIKY-UHFFFAOYSA-M
Synonym 1-allyl-3-methylimidazolium chloride, 1-allyl-3-methyl-1h-imidazol-3-ium chloride, 1-methyl-3-prop-2-en-1-yl imidazol-1-ium chloride, amim-cl, 1-allyl-3-methylimidazoliumchloride, dsstox_cid_27904, dsstox_rid_82657, dsstox_gsid_47928, 1-methyl-3-allylimidazolium chloride, 1-allyl-3-methyl-3-imidazolium chloride
PubChem CID 11321106
IUPAC Name 1-methyl-3-prop-2-enylimidazol-1-ium;chloride
SMILES C[N+]1=CN(C=C1)CC=C.[Cl-]
Specifications
Melting Point 49°C to 51°C
Quantity 5 g
Sensitivity Moisture sensitive
Formula Weight 158.63
Percent Purity 98%
Chemical Name or Material 1-Allyl-3-methylimidazolium chloride
compliance-icons
Product Identifier
  • 1-Allyl-3-methylimidazolium chloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : 1-Allyl-3-methylimidazolium chloride