accessibility menu, dialog, popup

UserName

CAS RN 65039-10-3

CH3NNCH2Cl

CAS RN 65039-10-3

IUPAC Name: 3-methyl-1-(prop-2-en-1-yl)-1H-imidazol-3-ium chloride
Synonyms: 1-methyl-3-(prop-2-en-1-yl)imidazol-1-ium chloride
Molecular Weight (g/mol): 158.63
Molecular Formula: C7H11ClN2
InChi Key: QVRCRKLLQYOIKY-UHFFFAOYSA-M
SMILES: [Cl-].C[N+]1=CN(CC=C)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results