missing translation for 'onlineSavingsMsg'
Learn More

1-Acetate 4-Methanesulfonate 1,4-Butanediol, TRC

Product Code. 30327926
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30327926 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30327926 Supplier Toronto Research Chemicals Supplier No. TRCA16940050MG

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-Acetate 4-Methanesulfonate 1,4-Butanediol
CAS 19859-00-8
Molecular Formula C7H14O5S
Quantity 50 mg
Synonym 4-((Methylsulfonyl)oxy)butyl Acetate,
InChI Formula InChI=1S/C7H14O5S/c1-7(8)11-5-3-4-6-12-13(2,9)10/h3-6H2,1-2H3
SMILES O=C(OCCCCOS(=O)(C)=O)C
IUPAC Name 4-methylsulfonyloxybutyl acetate
Molecular Weight (g/mol) 210.25
Formula Weight 210.0562
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.