missing translation for 'onlineSavingsMsg'
Learn More

1,9-Dichlorononane, TRC

Product Code. 30382956
Change view
Click to view available options
Quantity:
250 mg
1 g
2.5 g
Unit Size:
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30382956 1 g -
30381665 250 mg 250mg
30371435 2.5 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30382956 Supplier Toronto Research Chemicals Supplier No. TRCD4346251G
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 821-99-8
Molecular Formula C9H18Cl2
Molecular Weight (g/mol) 197.15
Synonym NSC 9477
IUPAC Name 1,9-dichlorononane
SMILES ClCCCCCCCCCCl

Specifications

Quantity 1 g
InChI Formula InChI=1S/C9H18Cl2/c10-8-6-4-2-1-3-5-7-9-11/h1-9H2
Formula Weight 196.0786
Chemical Name or Material 1,9-Dichlorononane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.