Learn More
1,8-Naphthalimide, 96%, BLD Pharm™
Specifications
Specifications
| CAS | 81-83-4 |
| Molecular Formula | C12H7NO2 |
| MDL Number | MFCD00006920 |
| Quantity | 5 g |
| SMILES | O=C1NC(C2=CC=CC3=CC=CC1=C23)=O |
| Molecular Weight (g/mol) | 197.19 |
| Percent Purity | 96% |
| Chemical Name or Material | 1,8-Naphthalimide |