Learn More
1,8-Dinitroanthraquinone, TRC
Product Code. 30363185
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30363185 500 mg 500mg
30356482 100 mg 100mg
30355859 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30363185 Supplier Toronto Research Chemicals Supplier No. TRCD191835500MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 129-39-5
Molecular Formula C14H6N2O6
Molecular Weight (g/mol) 298.21
Synonym 1,8-Dinitro-9,10-anthracenedione, 1,8-Dinitro-9,10-anthraquinone, Alizarine Violet R Base, NSC 37108, Waxoline Violet BA
IUPAC Name 1,8-dinitroanthracene-9,10-dione
SMILES [O-][N+](=O)c1cccc2C(=O)c3cccc(c3C(=O)c12)[N+](=O)[O-]
Specifications
Quantity 500 mg
InChI Formula InChI=1S/C14H6N2O6/c17-13-7-3-1-5-9(15(19)20)11(7)14(18)12-8(13)4-2-6-10(12)16(21)22/h1-6H
Formula Weight 298.0226
Chemical Name or Material 1,8-Dinitroanthraquinone